missing translation for 'onlineSavingsMsg'
Learn More

SR-18292, MedChemExpress

Product Code. 16456117
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456117

missing translation for 'mfr': Medchem Express HY101491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR-18292 is a PPAR gamma coactivator-1α (PGC-1α) inhibitor, which increases PGC-1α acetylation, suppresses gluconeogenic gene expression and reduces glucose production in hepatocytes.

SR-18292 is a PPAR gamma coactivator-1α (PGC-1α) inhibitor, which increases PGC-1α acetylation, suppresses gluconeogenic gene expression and reduces glucose production in hepatocytes.

Specifications

Chemical Name or Material SR-18292
CAS 2095432-55-4
Color Dust Gray
Purity Grade Notes Research
Molecular Formula C23H30N2O2
Quantity 5 mg
Solubility Information Ethanol : 100 mg/mL (272.85 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (272.85 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(CN(C(C)(C)C)CC(O)COC2=CC=CC3=C2C=CN3)C=C1
Molecular Weight (g/mol) 366.5
Formula Weight 366.5
Percent Purity 98.0%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.