Learn More
SR-18292, MedChemExpress
Product Code. 16493388
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16493388
Brand: Medchem Express HY10149110mg
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress SR-18292 is a PPAR gamma coactivator-1α (PGC-1α) inhibitor, which increases PGC-1α acetylation, suppresses gluconeogenic gene expression and reduces glucose production in hepatocytes.
SR-18292 is a PPAR gamma coactivator-1α (PGC-1α) inhibitor, which increases PGC-1α acetylation, suppresses gluconeogenic gene expression and reduces glucose production in hepatocytes.
Specifications
Chemical Name or Material SR-18292
CAS 2095432-55-4
Color Dust Gray
Purity Grade Notes Research
Molecular Formula C23H30N2O2
Quantity 10 mg
Solubility Information Ethanol : 100 mg/mL (272.85 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (272.85 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(CN(C(C)(C)C)CC(O)COC2=CC=CC3=C2C=CN3)C=C1
Molecular Weight (g/mol) 366.5
Formula Weight 366.5
Percent Purity 98.0%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-sugar/lipid metabolism