missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPRR4 Monoclonal Antibody (1G5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524528
This item is not returnable.
View return policy
Description
Peptide Sequence: MSSQQQQRQQ QQCPPQRAQQ QQVKQPCQPP PVKCQETCAP KTKDPCAPQV KKQCPPKGTI IPAQQKCPSA QQASKSKQK.
Cross-linked envelope protein of keratinocytes. Involved in UV-induced cornification.
Specifications
| SPRR4 | |
| Monoclonal | |
| Unconjugated | |
| SPRR4 | |
| small proline rich protein 4; small proline-rich protein 4; SPRR4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 163778 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1G5 | |
| PBS with no preservative; pH 7.4 | |
| Q96PI1 | |
| SPRR4 | |
| SPRR4 (NP_775103.1, 1 a.a. ~ 79 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction