Learn More
Edmund Bühler GmbH™ Spring Clamp
Fasten Erlenmeyer flasks on the universal tray.
Brand: Edmund Bühler GmbH™ 0009 649
Additional Details : Weight : 1.00000kg
Description
- Stainless steel construction
- For fastening Erlenmeyer flasks on the universal tray
- The sizes are related to Erlenmeyer flasks, but are also suitable for round-bottom flasks, beakers, etc
Specifications
2000 mL | |
Spring Clamp | |
Stainless Steel |