Learn More
Edmund Bühler GmbH™ Spring Clamp
Product Code. 10009360
missing translation for 'orderingAttributeHoverText'
Holds:
10 mL Flask
100 mL
1000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
missing translation for 'unitSize'
Each
Product Code. 10009360
missing translation for 'mfr': Edmund Bühler GmbH™ 0009 643
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fasten Erlenmeyer flasks on the universal tray.
  • Stainless steel construction
  • For fastening Erlenmeyer flasks on the universal tray
  • The sizes are related to Erlenmeyer flasks, but are also suitable for round-bottom flasks, beakers, etc
Specifications
Holds 25 mL
Product Type Spring Clamp
Material Stainless Steel