Learn More
Uvex Sport Blue PU 65999 Low Shoe - Width 12
Offers first-class wearer comfort with a sporty design; low weight, climate-optimizes high-tech materials and ergonomic outsole
125.00 €
Specifications
Color | Blue |
---|---|
Material | Textile |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17273171
|
Uvex
6599935 |
35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17283171
|
Uvex
6599936 |
36 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17293171
|
Uvex
6599937 |
37 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17203181
|
Uvex
6599938 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17213181
|
Uvex
6599939 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17223181
|
Uvex
6599940 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17233181
|
Uvex
6599941 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17243181
|
Uvex
6599942 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17253181
|
Uvex
6599943 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17263181
|
Uvex
6599944 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17273181
|
Uvex
6599945 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17283181
|
Uvex
6599946 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17293181
|
Uvex
6599947 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17203191
|
Uvex
6599948 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17213191
|
Uvex
6599949 |
49 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17223191
|
Uvex
6599950 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17233191
|
Uvex
6599951 |
51 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17243191
|
Uvex
6599952 |
52 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Beschreibung
Uvex 1 Sport Shoe 6599.9 S1 SRC
- Modern, ultra-lightweight and flexible S1 safety shoe
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials are free of silicones plasticizers and other paint wetting impairment substances
- Meets ESD specification with leakage resistance less than 35 megohms
- 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good lateral stability, no thermal conductivity
- Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
- Made with new developed technology for exceptional wearer comfort, climate-optimizes and breathable materials
- Virtually seam-free construction made from high-tech materials to eliminate pressure points
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Soft, padding on collar and tongue
- For light applications
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6599.9
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 35 to 52
Spezifikation
Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Textile | |
Composite | |
EN ISO 20345:2011 | |
Wide width | |
Yes | |
S1 |