Learn More
Uvex Sport Blue PU 65997 Low Shoe - Width 10
Offers first-class wearer comfort with a sporty design; low weight, climate-optimizes high-tech materials and ergonomic outsole
125.00 €
Specifications
Color | Blue |
---|---|
Material | Textile |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Composite |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17213141
|
Uvex
6599735 |
35 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17223141
|
Uvex
6599736 |
36 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17233141
|
Uvex
6599737 |
37 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17243141
|
Uvex
6599738 |
38 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17253141
|
Uvex
6599739 |
39 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17263141
|
Uvex
6599740 |
40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17273141
|
Uvex
6599741 |
41 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17283141
|
Uvex
6599742 |
42 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17293141
|
Uvex
6599743 |
43 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17203151
|
Uvex
6599744 |
44 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17213151
|
Uvex
6599745 |
45 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17223151
|
Uvex
6599746 |
46 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17233151
|
Uvex
6599747 |
47 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17243151
|
Uvex
6599748 |
48 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17253151
|
Uvex
6599749 |
49 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17263151
|
Uvex
6599750 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17273151
|
Uvex
6599751 |
51 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17283151
|
Uvex
6599752 |
52 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Uvex 1 Sport Shoe 6599.7 S1 SRC
- Modern, ultra-lightweight and flexible S1 safety shoe
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials are free of silicones plasticizers and other paint wetting impairment substances
- Meets ESD specification with leakage resistance less than 35 megohms
- 100% metal-free Uvex Xenova™ toe cap
- Compact design, anatomical shape, good lateral stability, no thermal conductivity
- Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
- Made with new developed technology for exceptional wearer comfort, climate-optimizes and breathable materials
- Virtually seam-free construction made from high-tech materials to eliminate pressure points
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Soft, padding on collar and tongue
- For light applications
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 6599.7
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 35 to 52
Specifications
Blue | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 1 | |
No | |
SRC |
Textile | |
Composite | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S1 |