Learn More
SPOP Antibody (S01-5B7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19781-100ul
This item is not returnable.
View return policy
Description
SPOP Monoclonal antibody specifically detects SPOP in Human samples. It is validated for Western Blot
Specifications
| SPOP | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8405 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S01-5B7 | |
| Unconjugated | |
| HIB homolog 1, Roadkill homolog 1, speckle-type POZ protein, TEF2 | |
| A synthetic peptide of human SPOP (Uniprot # O43791) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |