Learn More
STEMCELL Technologies Spleen Dissociation Medium
Product Code. 17159911
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 4 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17159911 10 x 4 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17159911 Supplier STEMCELL Technologies Supplier No. 07915
Please to purchase this item. Need a web account? Register with us today!
Medium for dissociation of mouse spleen
  • This product, containing enzymes, is designed to maximize the viability and recovery of dendritic cells from mouse spleen when used with EasySep™ cell separation technology
  • A single 4 mL tube is sufficient for processing up to two mouse spleens
Specifications
Cell Type Dendritic Cells
Quantity 10 x 4 mL