Learn More
STEMCELL Technologies Spleen Dissociation Medium
Description
- This product, containing enzymes, is designed to maximize the viability and recovery of dendritic cells from mouse spleen when used with EasySep™ cell separation technology
- A single 4 mL tube is sufficient for processing up to two mouse spleens
Specifications
Specifications
| Cell Type | Dendritic Cells |
| Quantity | 10 x 4 mL |