Learn More
Spiro[cyclopropane-1,2'-indan]-1'-one, TRC
Product Code. 30416856
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416856 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30416856 Supplier Toronto Research Chemicals Supplier No. TRCS682985500MG
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Spiro[cyclopropane-1,2'-indan]-1'-one
CAS 22228-23-5
Molecular Formula C11H10O
Quantity 500 mg
Synonym Spiro[cyclopropane-1,2'-[2H]inden]-1'(3'H)-one
InChI Formula InChI=1S/C11H10O/c12-10-9-4-2-1-3-8(9)7-11(10)5-6-11/h1-4H,5-7H2
SMILES O=C1c2ccccc2CC13CC3
IUPAC Name spiro[3H-indene-2,1'-cyclopropane]-1-one
Molecular Weight (g/mol) 158.2
Formula Weight 158.0732