Learn More
Integra Biosciences™ Spinner Bottles
Product Code. 10378901
Encompass
Change view
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10378901 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10378901 Supplier Integra Biosciences™ Supplier No. 182101
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The CELLSPIN is a moisture-resistant stirrer for cultivating cells in spinner bottles with culture volumes from 100mL to 1L. It offers a wide range of stirring options for accurate optimisation and control of culture conditions. The speed (5rpm to 75rpm) and other stirring parameters can be individually programmed and modified at any time via the control unit, which is located outside of the incubator. In case of power failure or accidental disconnection of control and spinner units an alarm system is activated.
  • The stirring unit is suitable for four flask sizes (100mL, 250mL, 500mL, 1L)
  • The medium oxygenation is improved and detrimental shear forces are minimised owing to the special design of the spinner flasks
  • Durable: no damage resulting from the incubator moisture owing to the sealed stirring unit and the removable control unit
  • The cultivation success is maximised thanks to a wide range of stirring options
  • Efficient documentation is possible through automatic data recording
  • Process security is increased by the instrument failure alerting function For accurate optimisation of cell cultivation in spinner bottles.
Specifications
Capacity (Metric) 1000 mL
For Use With (Equipment) For accurate optimization of cell cultivation in spinner bottles.