missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike RBD Antibody (1038345) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB107073JF669
This item is not returnable.
View return policy
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in MERS-CoV samples. It is validated for Flow Cytometry
Specifications
| Spike RBD | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 1038345 | |
| 50mM Sodium Borate | |
| Chinese Hamster Ovary cell line CHO-derived MERS-CoV Spike RBD, Glu367-Tyr606, Accession # YP_007188579.1 | |
| RUO | |
| MERS-CoV | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction