missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPHK1 Recombinant Human Protein
SPHK1, along with SPHK2, catalyzes the phosphorylation of sphingosine producing the signalling molecule sphingosine-1-phosphate (S1P).
1826.00 € - 7890.00 €
Specifications
Accession Number | NP_068807.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 71.4kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428693
|
Invitrogen™
PR8902A |
100μg |
1826.00 €
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418783
|
Invitrogen™
PV5215 |
1mg |
7890.00 €
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
S1P signals through G-protein coupled receptors and controls diverse cellular processes. Inhibitors of SPHKs are viewed as potential therapuetics for the treatment of cancer.
Specifications
NP_068807.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
SPHK and DGCK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
71.4kDa | |
RUO - research use only | |
SPHK1 | |
Human | |
GST Tag | |
Frozen |