Learn More
Cole-Parmer™ Mid-Size Vial Adapter for Extractor 04576-38
Shop All SPEX Certiprep Products
Click to view available options
For Use With (Equipment):
6804 Large Extractor/Vial Opener (04576-38)
Unit Size:
Each
Beschreibung
- Converts Model 6804 for use with 6881, 6883 and 6885 grinding vial sets.
Spezifikation
Spezifikation
| Product Type | Adapter |
| For Use With (Application) | Converts 6804 for Use with 6881, 6883 and 6885 Grinding Vial Sets |
| For Use With (Equipment) | 6804 Large Extractor/Vial Opener (04576-38) |