Learn More
SPEX™ SamplePrep Cryo-Adapter for 2210 Titer Plates
Extruded aluminum insert for 2210 96-well titer plate with round wells
Brand: SPEX™ SamplePrep 2650
Additional Details : Weight : 5.00000kg
Description
- Cryo-Blocks are chilled in the 2600 Cryo-Station and then used for high-throughput cryogenic grinding
- Enables high-throughput cryogenic grinding in the Geno/Grinder™
- Maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis
- Preserves pesticides, RNA, and proteins for heat sensitive extraction methods
- Increases yields by grinding samples at sub-zero temperatures
- Used two at a time in the Standard Clamp Assembly to keep it in balance
Specifications
Cryo Adapter | |
Ideal for High Throughput Cryogenic Grinding | |
2210 Titer Plate | |
Aluminum |