Learn More
Motic™ Specimen Holder, 2 Slides
Product Code. 15425252
Click to view available options
For Use With (Equipment):
BA310E Microscopes
Unit Size:
Each
Product Code. 15425252
Brand: Motic™ 1101001904131
Only null left
Add to basket
Add to basket
Hold 2 slide preparations for convenient change and rapid comparison. Suitable for use with BA310E Microscopes.
  • Holds two slides.
  • Facilitates rapid and simple multiple slide examination.
  • Increases ease of rapid slide comparison.
Specifications
Type Specimen Holder
Device Holds 2 Slides
For Use With (Equipment) BA310E Microscopes