missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specially Denatured Alcohol (SDA) 40B 190 Proof, Greenfield
Click to view available options
:
Fluorinated Poly Gallon
Specifications
Specifications
| CAS | Ethanol 64-17-5 62 - < 100 |
| Boiling Point | > 78.3 - < 80°C |
| Quantity | 4x1 Fluorinated Poly Gallon Case |
| Packaging | 4x1 Fluorinated Poly Gallon Case |
| Chemical Name or Material | Specially Denatured Alcohol (SDA) 40B 190 Proof |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction