Learn More
Spastazoline, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Spastazoline is a selective spastin (a microtubule-severing AAA protein) inhibitor, with an IC50 of 99 nM for Human spastin. Spastazoline can be used for cell division- and intracellular vesicle transport-related research.
Specifications
Specifications
| Chemical Name or Material | Spastazoline |
| Physical Form | Solid |
| CAS | 2351882-11-4 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H30N8 |
| Solubility Information | DMSO : 100 mg/mL (261.43 mM; Need ultrasonic) |
| SMILES | CC(C)(C)C1=CC(NC2=NC(N3CCN[C@H](C(C)C)C3)=NC4=C2C=CN4)=NN1 |
| Molecular Weight (g/mol) | 382.51 |
| Show More |