missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP3/SP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538652
This item is not returnable.
View return policy
Description
This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. A related pseudogene has been identified on chromosome 13. [provided by RefSeq, Feb 2010].
Specifications
| SP3/SP4 | |
| Polyclonal | |
| Unconjugated | |
| SP3 | |
| DKFZp686O1631, SPR2, Transcription factor Sp3, Transcription factor Sp4, SPR1, SPR-1, SPR-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20687, 20688, 25162, 367846, 6670, 6671 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q02446, Q02447, O70494, Q62445 | |
| SP3, SP4 | |
| A synthetic peptide derived from the C-terminal region of human SP3/4 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction