missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540681
This item is not returnable.
View return policy
Description
Peptide sequence: MVQQTNNAEN TEALLAGESS DSGAGLELGI ASSPTPGSTA STGGKADDPS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%.
SOX genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. SOX genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of SOX genes cluster at a minimum of 40 different loci that rapidly diverged in various animal lineages. At present 30 SOX genes have been identified, and members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
Specifications
| SOX4 | |
| Polyclonal | |
| Unconjugated | |
| SOX4 | |
| AA682046; ecotropic viral integration site 16; EVI16; SOX4; Sox-4; SRY (sex determining region Y)-box 4; SRY box 4; SRY-box 4; SRY-box containing gene 4; SRY-box transcription factor 4; SRY-related HMG-box gene 4; transcription factor SOX-4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6659 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q06945 | |
| SOX4 | |
| Synthetic peptide directed towards the N-terminal of human SOX4 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction