missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Mouse anti-Human, Clone: 1D2C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66786-1-IG-20UL
This item is not returnable.
View return policy
Description
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.Specifications
| SOX10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P56693 | |
| SOX10 | |
| SOX10 Fusion Protein Ag23191 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1D2C8 | |
| Unconjugated | |
| SOX10 | |
| DOM, SOX10, Transcription factor SOX 10, WS2E, WS4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6663 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction