Learn More
Sotetsuflavone, MedChemExpress
Description
Sotetsuflavone is a potent inhibitor of DENV-NS5 RdRp (Dengue virus NS5 RNA-dependent RNA polymerase) with an IC50 of 0.16 uM, is the most active compound of this series .
Specifications
Specifications
| Chemical Name or Material | Sotetsuflavone |
| CAS | 2608-21-1 |
| Color | Brown |
| Purity Grade Notes | Research |
| Quantity | 1 mg |
| SMILES | O=C1C=C(C2=CC=C(O)C=C2)OC3=C(C4=CC(C5=CC(C6=C(O)C=C(O)C=C6O5)=O)=CC=C4O)C(OC)=CC(O)=C13 |
| Molecular Weight (g/mol) | 552.48 |
| Grade | Research |
| Physical Form | Solid |