Learn More
Invitrogen™ Somatostatin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585759
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Pig (100%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%). Positive Control: adult mouse cerebellum. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.
Specifications
| Somatostatin | |
| Polyclonal | |
| Unconjugated | |
| SST | |
| Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20604, 24797, 6750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.71 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P60041, P60042, P61278 | |
| SST | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of mouse Somatostatin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.