Learn More
BUCHI Solid Loader M Set
Product Code. 17853360 Shop All Buchi Products
Change view
Click to view available options
Type:
Empty Solid Loader
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
17853360 Empty Solid Loader Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17853360 Supplier BUCHI Supplier No. 11070505
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Solid loader set for max. 40 g sample/silica
  • Enhancie separation resolution using dry loading
  • Incl. adapter set, sleeve, tubes (20pcs) and frits (40pcs)
TRUSTED_SUSTAINABILITY
Spezifikation
Type Empty Solid Loader