Learn More
Brady™ Brass Padlocks
Product Code. 15683497
Click to view available options
Color:
Black
Blue
Gray
Green
Red
Yellow
Unit Size:
Pack of 12
Product Code. 15683497
Brand: Brady™ 051357
Only null left
Add to basket
Add to basket
Features solid brass body and shackle that resists corrosion and sparks.
  • 6-pin cylinder offers more unique key cuts for better tamper resistance
  • Locks are protected by coloured vinyl sheaths making it easy to color-code your lockout program
  • Padlocks are available keyed different
  • Lock labels sold separately
  • Two keys supplied with each padlock
Specifications
Product Type Brass Padlocks
Alternate Part Number 51357
Color Yellow
Material Brass
Description Brass Padlocks
Diameter (Metric) Shackle 6.5 mm
Dimensions (L x W) 43.00 x 60.00 mm
Height (Metric) 21 mm
Height (Metric) Shackle 21.00 mm
Material (Shackle) Brass
No. of Pins 6
Quantity 12 Pk.
Width (Metric) 33.5 mm