missing translation for 'onlineSavingsMsg'
Learn More

Solcitinib, MedChemExpress

Product Code. p-200099625
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499707

Brand: Medchem Express HY16755

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Solcitinib is an orally active, competitive, potent, selective JAK1 inhibitor, with an IC50 of 9.8 nM, and 11-, 55- and 23-fold selectivity over JAK2, JAK3 and TYK2, respectively; Solcitinib is used in the research of moderate-to-severe plaque-type psoriasis.

Solcitinib is an orally active, competitive, potent, selective JAK1 inhibitor, with an IC50 of 9.8 nM, and 11-, 55- and 23-fold selectivity over JAK2, JAK3 and TYK2, respectively

Specifications

Chemical Name or Material Solcitinib
CAS 1206163-45-2
Color White
Purity Grade Notes Research
Molecular Formula C22H23N5O2
Quantity 5 mg
Synonym GSK-2586184 GLPG-0778
Solubility Information DMSO : 20 mg/mL (51.35 mM; Need ultrasonic and warming)
SMILES O=C(C1CC1)NC2=NN3C(C4=CC=C(C(N5CC(C)(C)C5)=O)C=C4)=CC=CC3=N2
Molecular Weight (g/mol) 389.45
Formula Weight 389.45
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.