missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOGA1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560576
This item is not returnable.
View return policy
Description
Immunogen sequence: LPGLREQAAL VSKAIDVLVA DANGFTAGLR LCLDNECADF RLHEAPDNSE GPRDTKLIHA ILVRLSVLQQ ELNAFTRKAD AVLGCSVKEQ QESF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
Specifications
| SOGA1 | |
| Polyclonal | |
| Unconjugated | |
| Soga1 | |
| 80-kDa SOGA fragment; 9830001H06Rik; AI426038; C20orf117; C-terminal 80 kDa form; D430036N24Rik; dJ132F21.1; KIAA0889; N-terminal form; Protein SOGA1; Soga; SOGA family member 1; Soga1; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140710 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94964 | |
| Soga1 | |
| Recombinant protein corresponding to Human SOGA1. Recombinant protein control fragment (Product #RP-99438). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction