Learn More
Sodium Trimethylacetate Hydrate, TRC
Spezifikation
Spezifikation
| Chemical Name or Material | Sodium Trimethylacetate Hydrate |
| CAS | 143174-36-1 |
| Molecular Formula | C5H11NaO3 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C5H10O2.Na.H2O/c1-5(2,3)4(6)7;;/h1-3H3,(H,6,7);;1H2/q;+1;/p-1 |
| SMILES | O.[Na+].CC(C)(C)C(=O)[O-] |
| IUPAC Name | sodium;2,2-dimethylpropanoate;hydrate |
| Molecular Weight (g/mol) | 142.13 |
| Formula Weight | 142.0606 |