Learn More
Sodium Trifluoroacetate-13C, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Trifluoroacetate-1-13C |
| CAS | 286425-32-9 |
| Molecular Formula | 13C C F3 O2 . Na |
| Quantity | 100 mg |
| Synonym | Acetic-1-13C acid','2,2,2-trifluoro-','sodium salt (9CI), 2,2,2-Trifluoroacetic-1-13C acid sodium salt, Trifluoroacetic-1-13C acid sodium salt, Sodium 2,2,2-trifluoroacetate-1-13C, Sodium trifluoroacetate-1-13C |
| InChI Formula | InChI=1S/C2HF3O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1/i1+1; |
| SMILES | [Na+].[O-][13C](=O)C(F)(F)F |
| IUPAC Name | sodium;2,2,2-trifluoroacetate |
| Molecular Weight (g/mol) | 136.998 |
| Formula Weight | 136.978 |