missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate, Anhydrous, ≥98% (calc. to the dried substance), Honeywell

Product Code. 15681350 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 kg
Packaging:
Paper / Poly Bag
Unit Size:
25kg
This item is not returnable. View return policy

Product Code. 15681350

Brand: Honeywell Research Chemicals 1348125KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7772-98-7
Physical Form Anhydrous
Quantity 25 kg
Molecular Formula Na2O3S2
Linear Formula Na2S2O3
MDL Number MFCD00003499
UN Number NONH for all modes of transport
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium sulfanidesulfonate
Molecular Weight (g/mol) 158.10
PubChem CID 24477
Formula Weight 158.11g/mol
Percent Purity >98%
Packaging Paper / Poly Bag
Assay calc. to the dried substance
Chemical Name or Material Sodium thiosulfate, Anhydrous, ≥98% (calc. to the dried substance)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.