Learn More
Sodium Perfluoro-1-decanesulfonate, TRC
Product Code. 18062330
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18062330 5 mg 5mg
18093400 2.5 mg 2.5mg
18020400 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18062330 Supplier Toronto Research Chemicals Supplier No. TRCS0809505MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2806-15-7
Molecular Formula C10 F21 O3 S . Na
Molecular Weight (g/mol) 622.13
Synonym 1-Decanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heneicosafluoro-, sodium salt (9CI), Sodium perfluorodecanesulfonate
IUPAC Name sodium;1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosafluorodecane-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C10HF21O3S.Na/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)35(32,33)34;/h(H,32,33,34);/q;+1/p-1
Formula Weight 621.9131
Percent Purity >95%
Chemical Name or Material Perfluorodecanesulfonic acid sodium salt
Grade HPLC