Learn More
Sodium N-(4-Hydroxyphenyl)acetamide-2,2,2-d3 Sulfate, CDN
Specifications
Specifications
| Chemical Name or Material | Paracetamol O-Sulfate D3 (acetyl-D3) Sodium Salt |
| CAS | 2733160-80-8 |
| Molecular Formula | C8 D3 H5 N O5 S . Na |
| Quantity | 0.01 g |
| Synonym | N-[4-(Sulfooxy)phenyl]acetamide-2,2,2-d3 sodium salt, 4-Acetaminophen-d3 sulfate sodium salt, Sodium acetaminophen sulfate-d3, N-(Acetyl-d3)-4-aminophenol O-sulfate sodium salt, Paracetamol O-sulfate D3 (acetyl-D3) sodium salt, Sodium paracetamol-d3 sulfate |
| InChI Formula | InChI=1 S/C8H9NO5S.Na/c1-6(10)9-7-2-4-8(5-3-7)14-15(11,12)13;/h2-5 H,1H3,(H,9,10)(H,11,12,13);/q;+1/p-1/i1D3; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C(=O)Nc1ccc(OS(=O)(=O)[O-])cc1 |
| IUPAC Name | sodium;[4-[(2,2,2-trideuterioacetyl)amino]phenyl] sulfate |
| Molecular Weight (g/mol) | 256.226 |
| Formula Weight | 256.021 g/mol |
| Show More |