Learn More
Sodium Methanesulfonate-d3, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Methyl-d3-sulfinic acid sodium salt |
| CAS | 2129597-38-0 |
| Molecular Formula | C D3 O2 S . Na |
| Quantity | 50 mg |
| Synonym | Methane-d3-sulfinic acid','sodium salt (1:1) (ACI), Sodium methane-d3-sulfinate, Sodium methylsulfinate-d3, Methane-d3-sulfinic acid sodium salt, Sodium methanesulfinate-D3 |
| InChI Formula | InChI=1S/CH4O2S.Na/c1-4(2)3;/h1H3,(H,2,3);/q;+1/p-1/i1D3; |
| SMILES | [Na+].[2H]C([2H])([2H])S(=O)[O-] |
| IUPAC Name | sodium;trideuteriomethanesulfinate |
| Molecular Weight (g/mol) | 105.1066 |
| Formula Weight | 104.994 |