Learn More
Sodium Metatungstate Hydrate, TRC
Product Code. 30412049
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412049 25 g 25g
30409997 1 g 1g
30413505 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30412049 Supplier Toronto Research Chemicals Supplier No. TRCS66750025G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 12141-67-2
Molecular Formula O39Na6W12•H2O
Molecular Weight (g/mol) 2986.01
Synonym Sodium metatungstate (Na6H2W12O40), Sodium tungstate (Na6H2W12O40), Sodium tungstate(VI) (Na6H2W12O40), Tungstate (W12(OH)2O386-) Hexasodium, Tungstic Acid (H8W12O40) Hexasodium Salt, SPT, POM 1
IUPAC Name hexasodium;dihydroxy(dioxo)tungsten;dioxido(dioxo)tungsten;trioxotungsten
SMILES OC(=O)c1ccc(cc1)NC[C@@H]2N(C=O)C3=C(NC(N)=NC3=O)NC2
Specifications
Quantity 25 g
InChI Formula InChI=1S/6Na.2H2O.38O.12W/h;;;;;;2*1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/q6*+1;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;6*-1;;;;;;;;;;;;+2/p-2
Formula Weight 2987.162
Chemical Name or Material Sodium Metatungstate Hydrate