Learn More
Sodium Lactobionate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Lactobionate |
| CAS | 27297-39-8 |
| Molecular Formula | C12H22O12.Na |
| Quantity | 100 g |
| Synonym | Lactobionic Acid Monosodium Salt, 4-(β-D-Galactosido)-D-gluconic Acid Sodium Salt |
| InChI Formula | InChI=1S/C12H22O12.Na/c13-1-3(15)10(7(18)8(19)11(21)22)24-12-9(20)6(17)5(16)4(2-14)23-12;/h3-10,12-20H,1-2H2,(H,21,22);/q;+1/p-1/t3-,4-,5+,6+,7-,8-,9-,10-,12+;/m1./s1 |
| SMILES | O[C@H]([C@@H](O)C(O[Na])=O)[C@@H]([C@H](O)CO)O[C@@H]([C@@H]([C@H]1O)O)O[C@@H]([C@@H]1O)CO |
| Molecular Weight (g/mol) | 381.29 |