Learn More
Sodium L-Lactate-3,3,3-d3, CDN
Specifications
Specifications
| Chemical Name or Material | L-Lactic Acid-d3 Sodium Salt |
| CAS | 285979-84-2 |
| Molecular Formula | C3 2H3 H2 O3 . Na |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4 H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i1D3; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])[C@H](O)C(=O)[O-] |
| IUPAC Name | sodium;(2 S)-3,3,3-trideuterio-2-hydroxypropanoate |
| Molecular Weight (g/mol) | 115.08 |
| Formula Weight | 115.0325 g/mol |
| Percent Purity | 98 atom % D, min 96% Chemical Purity |