Learn More
Sodium Estrone-2,4,16,16-d4 Sulfate (Stabilized with TRIS, 50% w/w), TRC
Specifications
Specifications
| Chemical Name or Material | Estrone-2,4,16,16-d4 3-Sulfate sodium salt |
| CAS | 285979-80-8 |
| Molecular Formula | C18 2H4 H17 O5 S . Na |
| Quantity | 1 mg |
| Synonym | Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-(sulfooxy)-, sodium salt (1:1), Estra-1,3,5(10)-trien-17-one-2,4,16,16-d4, 3-(sulfooxy)-, sodium salt (9CI), Estrone-2,4,16,16-D4 3-sulfate sodium |
| InChI Formula | InChI=1S/C18H22O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3,5,10,14-16H,2,4,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,15-,16+,18+;/m1./s1/i3D,7D2,10D; |
| SMILES | [Na+].[2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])C4=O)[C@@H]3CCc2c([2H])c1OS(=O)(=O)[O-] |
| IUPAC Name | sodium;[(8R,9S,13S,14S)-2,4,16,16-tetradeuterio-13-methyl-17-oxo-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-3-yl] sulfate |
| Molecular Weight (g/mol) | 376.44 |
| Formula Weight | 376.1258 |