missing translation for 'onlineSavingsMsg'
Learn More

Sodium Difluoroacetate-13D2, TRC

Product Code. 30424875
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424875 1 mg 1mg
30415304 10 mg 10mg
30419966 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424875 Supplier Toronto Research Chemicals Supplier No. TRCS6550221MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Difluoroacetate-13C2
Molecular Formula 13C2 H F2 Na O2
Quantity 1 mg
Synonym Difluoroacetic Acid Sodium Salt-13C2
InChI Formula InChI=1S/C2H2F2O2.Na/c3-1(4)2(5)6;/h1H,(H,5,6);/q;+1/p-1/i1+1,2+1;
SMILES F[13CH](F)[13C](=O)O[Na]
IUPAC Name (2,2-difluoroacetyl)oxysodium
Molecular Weight (g/mol) 120
Formula Weight 119.991
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.