Learn More
Sodium Dehydrocholate, TRC
Specifications
Specifications
| Chemical Name or Material | Dehydrocholic acid sodium salt |
| CAS | 145-41-5 |
| Molecular Formula | C24 H33 O5 . Na |
| Quantity | 5 g |
| Synonym | 3,7,12-Trioxo-5β-cholan-24-oic acid sodium salt, Dilabil sodium |
| InChI Formula | InChI=1S/C24H34O5.Na/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26;/h13-14,16-18,22H,4-12H2,1-3H3,(H,28,29);/q;+1/p-1/t13-,14+,16-,17+,18+,22+,23+,24-;/m1./s1 |
| SMILES | [Na+].C[C@H](CCC(=O)[O-])[C@H]1CC[C@H]2[C@H]3[C@H](CC(=O)[C@]12C)[C@@]4(C)CCC(=O)C[C@H]4CC3=O |
| IUPAC Name | sodium;(4R)-4-[(5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3,7,12-trioxo-1,2,4,5,6,8,9,11,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]pentanoate |
| Molecular Weight (g/mol) | 424.5 |
| Formula Weight | 424.2226 |