Learn More
Sodium Colistin Methanesulfonate (Technical Grade), TRC
Product Code. 30428574
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428574 5 g 5g
30424216 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30428574 Supplier Toronto Research Chemicals Supplier No. TRCS6153005G
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 8068-28-8
Molecular Formula C58H105N16Na5O28S5
Molecular Weight (g/mol) 1749.8
Synonym Colistinmethanesulfonic Acid, Colimycin M, Colimycin Sodium Methanesulfonate, Colistimethate, Colistin Sodium Methanesulfonate, Colistin Sulfomethate, Colistin Sulfomethate Sodium, Colistin, Methyl Sulfate Sodium Salt, Colistinat, Coly-Mycin Injectable, Colymycin M, Promixin, Sodium Colistimethate, Sodium Colistinmethanesulfonate, W 1929
SMILES CC[C@@H](C)CCCCC(=O)N[C@@H](CCNCS(=O)(=O)[O-])C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCNCS(=O)(=O)[O-])C(=O)N[C@H]1CCNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](NC(=O)[C@@H](NC1=O)CCNCS(=O)(=O)[O-])CC(C)C)CC(C)C)CCNCS(=O)(=O)[O-])CCNCS(=O)(=O)[O-])[C@@H](C)O.[Na+].[Na+].[Na+].[Na+].[Na+]
Tekniske data
Quantity 5 g
InChI Formula InChI=1S/C58H110N16O28S5.5Na/c1-9-35(6)12-10-11-13-46(77)65-38(14-20-59-28-103(88,89)90)53(82)74-48(37(8)76)58(87)70-41(17-23-62-31-106(97,98)99)50(79)68-43-19-25-64-57(86)47(36(7)75)73-54(83)42(18-24-63-32-107(100,101)102)67-49(78)39(15-21-60-29-104(91,92)93)69-55(84)44(26-33(2)3)72-56(85)45(27-34(4)5)71-52(81)40(66-51(43)80)16-22-61-30-105(94,95)96;;;;;/h33-45,47-48,59-63,75-76H,9-32H2,1-8H3,(H,64,86)(H,65,77)(H,66,80)(H,67,78)(H,68,79)(H,69,84)(H,70,87)(H,71,81)(H,72,85)(H,73,83)(H,74,82)(H,88,89,90)(H,91,92,93)(H,94,95,96)(H,97,98,99)(H,100,101,102);;;;;/q;5*+1/p-5/t35-,36-,37-,38+,39+,40+,41+,42+,43+,44+,45-,47+,48+;;;;;/m1...../s1
Chemical Name or Material Colistimethate sodium
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.