missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chloride Solution, Volumetric, 0.5M NaCl (0.5N), Honeywell™ Fluka™

Product Code. 15656780 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15656780 1 L Poly bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15656780 Supplier Honeywell-Fluka Supplier No. 713931L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 7647-14-5
Quantity 1 L
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
UN Number NONH for all modes of transport
Beilstein 3534976
Synonym sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44g/mol
Packaging Poly bottle
Chemical Name or Material Sodium chloride solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.