Learn More
Sodium 4-pentynoate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Sodium 4-pentynoate is a alkynylacetate analogue, can be metabolically incorporated onto cellular proteins through biosynthetic mechanisms for profiling of acetylated proteins in diverse cell types.
Specifications
Specifications
| Chemical Name or Material | Sodium 4-pentynoate |
| Physical Form | Solid |
| CAS | 101917-30-0 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C5H5NaO2 |
| Solubility Information | H2O : 100 mg/mL (832.78 mM; Need ultrasonic) ∣DMSO : 16.67 mg/mL (138.82 mM; Need ultrasonic) |
| SMILES | C#CCCC(O[Na])=O |
| Molecular Weight (g/mol) | 120.08 |
| Show More |