Learn More
BRAND™ Soda Lime Glass Threaded Bottles
Shop All Brand Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2500 mL
Coating:
None
ethylene-acrylate coating
Unit Size:
Each
Description
- Soda-lime glass (amber)
- Screw-cap, polypropene (PP), pouring ring PE-LD
- Round glass narrow mouth bottles
- Space-saving square base
- Available ethylene-acrylate-coated or uncoated
- The coating envelops the glass bottle like a protection skin
- Should the bottle break, the splintering hazards are reduced considerably
- The maximum operating temperature for coated bottles is 80 °C
- To preserve the coating, do not clean at temperatures exceeding 60 ºC
- With PP screw cap GL45
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Coating | ethylene-acrylate coating |
| Color | Amber |
| Material | Soda Lime Glass |