Learn More
Brand™ Soda Lime Glass Culture Tubes
Product Code. 30243948 Shop All Brand Products
Click to view available options
Length (Metric):
75 mm
100 mm
125 mm
160 mm
Diameter (Metric):
10 mm
12 mm
16 mm
Unit Size:
Pack of 100
Pack of 105
Pack of 144
Pack of 250
Pack of 344
Pack of 78
Product Code. 30243948
Brand: Brand™ 114125
Please to purchase this item. Need a web account? Register with us today!
Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes are rimless and autoclavable at 121°C.
  • Manufactured from soda lime glass.
  • Rimless
  • Autoclavable at 121°C.
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 160 mm
Quantity 100/Pk.
Diameter (Metric) 16 mm
Product Type Culture Tube