Learn More
Soblidotin, MedChemExpress
Product Code. 16476128
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476128 10 mg 10mg
16453436 25 mg 25mg
16452267 50 mg 50mg
16421439 1 mg 1mg
16408737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16476128 Supplier Medchem Express Supplier No. HY14672
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Soblidotin (Auristatin PE) is a novel synthetic Dolastatin 10 derivative and inhibitor of tubulin polymerization.
Soblidotin (Auristatin PE) is a novel synthetic Dolastatin 10 derivative and inhibitor of tubulin polymerization.
Specifications
Chemical Name or Material Soblidotin
Physical Form Solid
CAS 149606-27-9
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C39H67N5O6
For Use With (Application) Cancer-Kinase/protease
Synonym Auristatin PE TZT-1027
Solubility Information DMSO : 100 mg/mL (142.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N[C@@H](C(C)C)C(N([C@@H]([C@H](CC)C)[C@H](OC)CC(N1CCC[C@@]1([H])[C@H](OC)[C@@H](C)C(NCCC2=CC=CC=C2)=O)=O)C)=O)[C@H](C(C)C)N(C)C
Molecular Weight (g/mol) 701.98
Formula Weight 701.98
Percent Purity 99.39%
Grade Research