missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX20 Monoclonal Antibody (4E9)

Description
Peptide Sequence: SGLSSNSSMT TRELQQYWQN QKCRWKHVKL LFEIASARIE ERKVSKFVMG KSRPGEMTYP GSRGETGTAP EPDPRCPRQS DM.
SNX20 interacts with the cytoplasmic domain of PSGL1 and cycles PSGL1 into endosomes.
Specifications
Specifications
| Antigen | SNX20 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4E9 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SNX20 |
| Gene Accession No. | Q7Z614 |
| Gene Alias | 9130017C17Rik; RGD1307787; selectin ligand interactor cytoplasmic 1; selectin ligand interactor cytoplasmic-1; selectin ligand-interactor cytoplasmic 1; SLIC1; SLIC-1; SNX20; sorting nexin 20; sorting nexin-20 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?