missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNRPC Polyclonal Antibody
GREENER_CHOICE

Product Code. 15780912 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15780912 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15780912 Supplier Invitrogen™ Supplier No. PA563136

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VKDYYQKWME EQAQSLIDKT TAAFQQGKIP PTPFSAPPPA GAMIPPP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

SNRPC is associated with snRNP U1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNRPC
Gene Accession No. P09234
Gene Alias small nuclear ribonucleoprotein polypeptide C; Snrp1c; Snrpc; U1 small nuclear ribonucleoprotein 1C; U1 small nuclear ribonucleoprotein C; U1 small nuclear RNP specific C; U1 snRNP C; U1 snRNP protein C; U1C; U1-C; U1C protein; Yhc1
Gene Symbols SNRPC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SNRPC. Recombinant protein control fragment (Product #RP-103217).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.