missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smoothened Antibody (1D9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00006608-M11
This item is not returnable.
View return policy
Description
Smoothened Monoclonal antibody specifically detects Smoothened in Human samples. It is validated for Western Blot, ELISA
Specifications
| Smoothened | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Gx, Protein Gx, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila) | |
| SMO (NP_005622.1, 653 a.a. ~ 787 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NLWLVEAEISPELQKRLGRKKKRRKRKKEVCPLAPPPELHPPAPAPSTIPRLPQLPRQKCLVAAGAWGAGDSCRQGAWTLVSNPFCPEPSPPQDPFLPSAPAPVAWAHGRRQGLGPIHSRTNLMDTELMDADSDF | |
| 0.1 mg | |
| GPCR, Growth and Development, Neuronal Cell Markers | |
| 6608 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 1D9 | |
| Western Blot, ELISA | |
| NP_005622 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction