missing translation for 'onlineSavingsMsg'
Learn More

SMIP004, MedChemExpress

Product Code. 16442827
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442827 50 mg 50mg
16496876 100 mg 100mg
16469297 5 mg 5mg
16446748 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16442827 Supplier Medchem Express Supplier No. HY15694

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SMIP004 is a SKP2 E3 ligase inhibitor, which downregulates SKP2 and to stabilise p27. SMIP004 is a cancer cell selective apoptosis inducer of human prostate cancer cells.

SMIP004 is a SKP2 E3 ligase inhibitor, which downregulates SKP2 and to stabilise p27. SMIP004 is a cancer cell selective apoptosis inducer of human prostate cancer cells.

Specifications

Chemical Name or Material SMIP004
Physical Form Solid
CAS 143360-00-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C13H19NO
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 100 mg/mL (487.09 mM)
SMILES CC1=C(NC(C)=O)C=CC(CCCC)=C1
Molecular Weight (g/mol) 205.3
Formula Weight 205.3
Percent Purity 98.66%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.