missing translation for 'onlineSavingsMsg'
Learn More

SMI-16a, MedChemExpress

Product Code. 16412816
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16412816

Brand: Medchem Express HY101947

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SMI-16a is a selective Pim kinase inhibitor with IC50 values of 0.15, 0.02 and 48 μM for Pim1, Pim2 and PC3 cells, respectively.

SMI-16a is a selective Pim kinase inhibitor with IC50 values of 0.15, 0.02 and 48 μM for Pim1, Pim2 and PC3 cells, respectively.

Specifications

Chemical Name or Material SMI-16a
CAS 587852-28-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H13NO3S
Quantity 25 mg
Synonym PIM1/2 Kinase Inhibitor VI
Solubility Information DMSO : ≥ 100 mg/mL (379.78 mM)
SMILES O=C(NC/1=O)SC1=C\C2=CC=C(OCCC)C=C2
Molecular Weight (g/mol) 263.31
Formula Weight 263.31
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.